Julolidine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

J0007-5G J0007-25G
TCJ0007-5GEA 151.68 CAD
TCJ0007-5G TCJ0007-25G
Julolidine ≥97.0% (by GC, titration analysis)
Julolidine

CAS Number: 479-59-4
Molecular Formula: C12H15N
Molecular Weight: 173.26
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 280
Melting point (°C): 37
Flash Point (°C): 113
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizine

Formula: C₁₂H₁₅N
Boiling Pt: 280 °C
Melting Pt: 37 °C
Flash Pt: 113 °C
MDL Number: MFCD00006917
CAS Number: 479-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d85a779c8195b775545e7a6e9241080357c9046&vid=2d85a779c8195b775545e7a6e9241080357c9046&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR