cis-Jasmone ≥92.0%
Supplier: TCI America
Synonyms:
(Z)-3-Methyl-2-(pent-2-en-1-yl)cyclopent-2-enone, cis-Jasmon
J0003-5ML
J0003-25ML
TCJ0003-005MLEA
34.76
CAD
TCJ0003-005ML
TCJ0003-025ML
cis-Jasmone ≥92.0%
cis-Jasmone
CAS Number: 488-10-8
MDL Number: MFCD00001402
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >92.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 128
Specific Gravity (20/20): 0.94
Other Product Names / Synonyms: 3-Methyl-2-(cis-2-pentenyl)-2-cyclopenten-1-one
Formula:
C₁₁H₁₆O Boiling Pt: 135 °C Density: 0.94 (20 °C) Flash Pt: 128 °C |
MDL Number:
MFCD00001402 CAS Number: 488-10-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...