Janus green B
Supplier: TCI America
J0002-25G
J0002-5G
TCJ0002-025GEA
244.9
CAD
TCJ0002-025G
TCJ0002-005G
Janus green B
Janus green B
CAS Number: 2869-83-2
MDL Number: MFCD00011758
Molecular Formula: C30H31ClN6
Molecular Weight: 511.07
Form: Crystal
Color: Deep Green
Formula:
C₃₀H₃₁ClN₆ |
MDL Number:
MFCD00011758 CAS Number: 2869-83-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...