(3-Iodophenyl)acetic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-(3-Iodophenyl)acetic acid

I0760-1G I0760-5G
TCI0760-1GEA 61.62 CAD
TCI0760-1G TCI0760-5G
(3-Iodophenyl)acetic acid ≥98.0%
(3-Iodophenyl)acetic acid

CAS Number: 1878-69-9
MDL Number: MFCD00046548
Molecular Formula: C8H7IO2
Molecular Weight: 262.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 131

Formula: (C₆H₄I)CH₂COOH
Melting Pt: 131 °C
MDL Number: MFCD00046548
CAS Number: 1878-69-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db5b9c0ee4b39aea53200b3fff3779d9ff324b2e&vid=db5b9c0ee4b39aea53200b3fff3779d9ff324b2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR