2-Iodo-m-xylene ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dimethyl-2-iodobenzene, 2-Iodo-1,3-dimethylbenzene, 2,6-Dimethyliodobenzene, 2,6-Dimethyl-1-iodobenzene

I0752-5G I0752-25G
TCI0752-5GEA 58.46 CAD
TCI0752-5G TCI0752-25G
2-Iodo-m-xylene ≥98.0%
2-Iodo-m-xylene

CAS Number: 608-28-6
MDL Number: MFCD00019012
Molecular Formula: C8H9I
Molecular Weight: 232.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Melting point (°C): 11
Flash Point (°C): 102
Specific Gravity (20/20): 1.65


Other Product Names / Synonyms: 2-Iodo-1,3-dimethylbenzene

Formula: C₈H₉I
Boiling Pt: 230 °C
Melting Pt: 11 °C
Density: 1.65 (20 °C)
Flash Pt: 102 °C
MDL Number: MFCD00019012
CAS Number: 608-28-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85352959cb55e2b493efe6555169acbb74077bf3&vid=85352959cb55e2b493efe6555169acbb74077bf3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR