3,5-Dimethyl-4-iodoisoxazole ≥98.0%
Supplier: TCI America
I0727-1G
I0727-5G
TCI0727-1GEA
51.19
CAD
TCI0727-1G
TCI0727-5G
3,5-Dimethyl-4-iodoisoxazole ≥98.0%
3,5-Dimethyl-4-iodoisoxazole
CAS Number: 10557-85-4
MDL Number: MFCD00173743
Molecular Formula: C5H6INO
Molecular Weight: 223.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Formula:
C₅H₆INO Melting Pt: 54 °C |
MDL Number:
MFCD00173743 CAS Number: 10557-85-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...