(2-Iodophenyl)acetic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(2-Iodophenyl)acetic acid

I0723-25G I0723-5G
TCI0723-25GEA 278.08 CAD
TCI0723-25G TCI0723-5G
(2-Iodophenyl)acetic acid ≥97.0% (by GC, titration analysis)
(2-Iodophenyl)acetic acid

CAS Number: 18698-96-9
MDL Number: MFCD00046546
Molecular Formula: C8H7IO2
Molecular Weight: 262.05
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 117

Formula: (C₆H₄I)CH₂COOH
MW: 262.05 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00046546
CAS Number: 18698-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163d00f684186d95f2889f4011eb06155ab10547&vid=163d00f684186d95f2889f4011eb06155ab10547&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR