Indole-4-carbaldehyd ≥98.0%

Supplier: TCI America

I0712-5G I0712-1G
TCI0712-5GEA 661.23 CAD
TCI0712-5G TCI0712-1G
Indole-4-carbaldehyd ≥98.0%
Indole-4-carbaldehyd

CAS Number: 1074-86-8
MDL Number: MFCD01632221
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 144


Other Product Names / Synonyms: 4-Formylindole

Formula: C₉H₇NO
Melting Pt: 144 °C
MDL Number: MFCD01632221
CAS Number: 1074-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6598f0cd0b8417b0187f268bd0681924fdbe46ee&vid=6598f0cd0b8417b0187f268bd0681924fdbe46ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR