3-Iodobenzylamine hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

I0710-1G I0710-5G
TCI0710-1GEA 45.82 CAD
TCI0710-1G TCI0710-5G
3-Iodobenzylamine hydrochloride ≥98.0% (by HPLC)
3-Iodobenzylamine hydrochloride

CAS Number: 3718-88-5
MDL Number: MFCD00012857
Molecular Formula: C7H8IN
Molecular Weight: 269.51
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 194

Formula: C₇H₉ClIN
MW: 269.5105 g/mol
Melting Pt: 194 °C
MDL Number: MFCD00012857
CAS Number: 3718-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42b226e25dc2b9b09a03eba1fdf38d26da60393c&vid=42b226e25dc2b9b09a03eba1fdf38d26da60393c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR