3-Iodo-p-toluic acid ≥98.0%
Supplier: TCI America
I0707-25G
I0707-5G
TCI0707-25GEA
464.52
CAD
TCI0707-25G
TCI0707-5G
3-Iodo-p-toluic acid ≥98.0%
3-Iodo-p-toluic acid
CAS Number: 82998-57-0
MDL Number: MFCD00010392
Molecular Formula: C8H7IO2
Molecular Weight: 262.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 209
Other Product Names / Synonyms: 3-Iodo-4-methylbenzoic Acid
Formula:
C₈H₇IO₂ Melting Pt: 209 °C |
MDL Number:
MFCD00010392 CAS Number: 82998-57-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...