2-Iodo-4-nitrotoluene ≥97.0%
Supplier: TCI America
I0706-5G
I0706-25G
TCI0706-5GEA
66.04
CAD
TCI0706-5G
TCI0706-25G
2-Iodo-4-nitrotoluene ≥97.0%
2-Iodo-4-nitrotoluene
CAS Number: 7745-92-8
MDL Number: MFCD00024329
Molecular Formula: C7H6INO2
Molecular Weight: 263.03
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 165
Melting point (°C): 52
Formula:
C₇H₆INO₂ Boiling Pt: 165 °C Melting Pt: 52 °C |
MDL Number:
MFCD00024329 CAS Number: 7745-92-8 UN: 3077 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...