4-Iodo-o-toluidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0690-25G I0690-5G
TCI0690-25GEA 151.68 CAD
TCI0690-25G TCI0690-5G
4-Iodo-o-toluidine ≥98.0% (by GC, titration analysis)
4-Iodo-o-toluidine

CAS Number: 13194-68-8
MDL Number: MFCD00025299
Molecular Formula: C7H8IN
Molecular Weight: 233.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 88


Other Product Names / Synonyms: 2-Amino-5-iodotoluene, 4-Iodo-o-toluidine

Formula: C₇H₈IN
Melting Pt: 88 °C
MDL Number: MFCD00025299
CAS Number: 13194-68-8
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f37e83a3e924269f9df1a93ad74ac68f63b6b501&vid=f37e83a3e924269f9df1a93ad74ac68f63b6b501&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR