Indole-5-carbaldehyd ≥98.0%

Supplier: TCI America

I0674-25G I0674-5G
TCI0674-25GEA 925.88 CAD
TCI0674-25G TCI0674-5G
Indole-5-carbaldehyd ≥98.0%
Indole-5-carbaldehyd

CAS Number: 1196-69-6
MDL Number: MFCD02093664
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: 5-Formylindole

Formula: C₉H₇NO
Melting Pt: 100 °C
MDL Number: MFCD02093664
CAS Number: 1196-69-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c7d20686ccd879e0946a2e05ebcac310b7f925b&vid=7c7d20686ccd879e0946a2e05ebcac310b7f925b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR