4-Iodopyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0673-25G I0673-5G
TCI0673-25GEA 723.64 CAD
TCI0673-25G TCI0673-5G
4-Iodopyridine ≥98.0% (by GC, titration analysis)
4-Iodopyridine

CAS Number: 15854-87-2
MDL Number: MFCD02093937
Molecular Formula: C5H4IN
Molecular Weight: 205.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 108
Storage Temperature: <0°C

Formula: C₅H₄IN
MW: 205 g/mol
Melting Pt: 108 °C
MDL Number: MFCD02093937
CAS Number: 15854-87-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b86239555fb2343536ffbc847a34de36c2d6957&vid=9b86239555fb2343536ffbc847a34de36c2d6957&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR