1-Isoquinolinecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I0671-25G I0671-5G
TCI0671-25GEA 278.08 CAD
TCI0671-25G TCI0671-5G
1-Isoquinolinecarboxylic acid ≥98.0% (by GC, titration analysis)
1-Isoquinolinecarboxylic acid

CAS Number: 486-73-7
MDL Number: MFCD00006901
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 156

Formula: C₁₀H₇NO₂
Melting Pt: 156 °C
MDL Number: MFCD00006901
CAS Number: 486-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e4a0af4765113c0c8fbbb9ea81cc9f4ab49697b&vid=9e4a0af4765113c0c8fbbb9ea81cc9f4ab49697b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR