Indole-6-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0663-5G I0663-1G
TCI0663-5GEA 244.9 CAD
TCI0663-5G TCI0663-1G
Indole-6-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Indole-6-carboxylic acid

CAS Number: 1670-82-2
MDL Number: MFCD00210441
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 257

Formula: C₉H₇NO₂
Melting Pt: 257 °C
MDL Number: MFCD00210441
CAS Number: 1670-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76a69f9505b7b1a4da88f64de9bddea0c65a913f&vid=76a69f9505b7b1a4da88f64de9bddea0c65a913f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR