2-Indanone ≥98.0%

Supplier: TCI America

Synonyms: beta-Hydrindone, 2-Oxoindane, 1H-Inden-2(3H)-one

I0649-25G I0649-5G
TCI0649-25GEA 144.62 CAD
TCI0649-25G TCI0649-5G
2-Indanone ≥98.0%
2-Indanone

CAS Number: 615-13-4
MDL Number: MFCD00003792
Molecular Formula: C9H8O
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 57
Flash Point (°C): 100
Storage Temperature: 0-10°C


Other Product Names / Synonyms: beta-Hydrindone, 2-Oxoindane

Formula: C₉H₈O
Boiling Pt: 225 °C
Melting Pt: 57 °C
Flash Pt: 100 °C
MDL Number: MFCD00003792
CAS Number: 615-13-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=215cffa254a48fc822b9fd240434bbda83ecde41&vid=215cffa254a48fc822b9fd240434bbda83ecde41&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR