(4-Iodophenoxy)acetic acid ≥97.0%

Supplier: TCI America

I0644-25G I0644-5G
TCI0644-25GEA 244.9 CAD
TCI0644-25G TCI0644-5G
(4-Iodophenoxy)acetic acid ≥97.0%
(4-Iodophenoxy)acetic acid

CAS Number: 1878-94-0
Molecular Formula: C8H7IO3
Molecular Weight: 278.05
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 157

Formula: (C₆H₄I)OCH₂COOH
Melting Pt: 157 °C
MDL Number: MFCD00014359
CAS Number: 1878-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ba31e5df89af7318a5d7f99fe4969488782b8ba&vid=5ba31e5df89af7318a5d7f99fe4969488782b8ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR