Imidazole-4-carboxaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 1H-Imidazole-5-carboxaldehyde, 4-Formylimidazole, Imidazole-4-carbaldehyde

I0636-1G I0636-5G
TCI0636-1GEA 50.56 CAD
TCI0636-1G TCI0636-5G
Imidazole-4-carboxaldehyde ≥98.0%
Imidazole-4-carboxaldehyde

CAS Number: 3034-50-2
Molecular Formula: C4H4N2O
Molecular Weight: 96.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 175
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4-Formylimidazole

Formula: C₄H₄N₂O
MW: 96.09 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00173726
CAS Number: 3034-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dddb0377eaa0e5c1c1eabc78c269f3930b42e65a&vid=dddb0377eaa0e5c1c1eabc78c269f3930b42e65a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR