4-Isopropylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

I0620-5G I0620-1G
TCI0620-5GEA 191.81 CAD
TCI0620-5G TCI0620-1G
4-Isopropylphenylboronic acid (contains varying amounts of Anhydride)
4-Isopropylphenylboronic acid

CAS Number: 16152-51-5
MDL Number: MFCD01074614
Molecular Formula: C9H13BO2
Molecular Weight: 164.01
Form: Crystal
Color: White
Melting point (°C): 140


Other Product Names / Synonyms: 4-Isopropylbenzeneboronic Acid

Formula: C₉H₁₃BO₂
Melting Pt: 140 °C
MDL Number: MFCD01074614
CAS Number: 16152-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fdf681714fff0987622ad9179be68c9164e68b6&vid=7fdf681714fff0987622ad9179be68c9164e68b6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR