Iridium(III) chloride hydrate
Supplier: TCI America
Synonyms:
Iridium trichloride hydrate, Iridium(III) chloride xhydrate
I0616-1G
I0616-5G
TCI0616-1GEA
477.33
CAD
TCI0616-1G
TCI0616-5G
Iridium(III) chloride hydrate
Iridium (III) chloride hydrate
CAS Number: 14996-61-3
MDL Number: MFCD00149749
Molecular Formula: Cl3Ir
Molecular Weight: 298.57
Form: Crystal
Color: Deep Yellow
Melting point (°C): 763
Formula:
Cl₃Ir·xH₂O MW: 298.58 g/mol (anhy) Melting Pt: 763 °C |
MDL Number:
MFCD00149750 CAS Number: 14996-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...