4-Iodobenzaldehyde ≥96.0%

Supplier: TCI America

I0612-1G I0612-5G
TCI0612-1GEA 105.86 CAD
TCI0612-1G TCI0612-5G
4-Iodobenzaldehyde ≥96.0%
4-Iodobenzaldehyde

CAS Number: 15164-44-0
MDL Number: MFCD00039576
Molecular Formula: C7H5IO
Molecular Weight: 232.02
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 265
Melting point (°C): 77

Formula: C₇H₅IO
MW: 232.02 g/mol
Boiling Pt: 265 °C
Melting Pt: 77 °C
MDL Number: MFCD00039576
CAS Number: 15164-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f4034650d0260f2dc4225bb20ad761fab1a5039&vid=5f4034650d0260f2dc4225bb20ad761fab1a5039&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR