3-Iodobenzaldehyde ≥96.0%

Supplier: TCI America

I0611-25G I0611-5G
TCI0611-25GEA 439.24 CAD
TCI0611-25G TCI0611-5G
3-Iodobenzaldehyde ≥96.0%
3-Iodobenzaldehyde

CAS Number: 696-41-3
MDL Number: MFCD00039573
Molecular Formula: C7H5IO
Molecular Weight: 232.02
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 59

Formula: C₇H₅IO
Boiling Pt: 125 °C
Melting Pt: 59 °C
MDL Number: MFCD00039573
CAS Number: 696-41-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6c7cd53caaf4be8ee48e6627e591f2bd9193a0f&vid=a6c7cd53caaf4be8ee48e6627e591f2bd9193a0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR