Isobutylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

I0597-25G I0597-5G
TCI0597-25GEA 469.26 CAD
TCI0597-25G TCI0597-5G
Isobutylboronic acid (contains varying amounts of Anhydride)
Isobutylboronic acid

CAS Number: 84110-40-7
MDL Number: MFCD00134156
Molecular Formula: C4H11BO2
Molecular Weight: 101.94
Form: Crystal
Color: White
Melting point (°C): 104


Other Product Names / Synonyms: 1-Isobutaneboronic Acid

Formula: C₄H₁₁BO₂
Melting Pt: 104 °C
MDL Number: MFCD00134156
CAS Number: 84110-40-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bc6cc28ff1c940de5b8746fe7157aa490e8c433&vid=4bc6cc28ff1c940de5b8746fe7157aa490e8c433&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR