Ethyl indole-2-carboxylate ≥98.0%

Supplier: TCI America

I0585-25G I0585-5G
TCI0585-25GEA 1338.89 CAD
TCI0585-25G TCI0585-5G
Ethyl indole-2-carboxylate ≥98.0%
Ethyl indole-2-carboxylate

CAS Number: 3770-50-1
MDL Number: MFCD00005609
Molecular Formula: C11H11NO2
Molecular Weight: 189.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 122


Other Product Names / Synonyms: Indole-2-carboxylic Acid Ethyl Ester

Formula: C₁₁H₁₁NO₂
Melting Pt: 122 °C
MDL Number: MFCD00005609
CAS Number: 3770-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd7736f5cb5c778626f2919dd461e7fa05217f18&vid=fd7736f5cb5c778626f2919dd461e7fa05217f18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR