(R)-(+)-4-Isopropyl-2-oxazolidinone ≥98.0%
Supplier: TCI America
Synonyms:
(R)-4-Isopropyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-2-oxazolidinone
I0572-1G
I0572-25G
I0572-5G
TCI0572-1GEA
80.58
CAD
TCI0572-1G
TCI0572-25G
TCI0572-5G
(R)-(+)-4-Isopropyl-2-oxazolidinone ≥98.0%
(R)-(+)-4-Isopropyl-2-oxazolidinone
CAS Number: 95530-58-8
MDL Number: MFCD00075172
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72
Specific rotation [a]20/D: 18 deg (C=6, EtOH)
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Melting Pt: 72 °C |
MDL Number:
MFCD00075172 CAS Number: 95530-58-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...