4'-Isopropylacetophenone ≥95.0%

Supplier: TCI America

I0570-25ML I0570-5ML
TCI0570-25MLEA 330.22 CAD
TCI0570-25ML TCI0570-5ML
4'-Isopropylacetophenone ≥95.0%
4'-Isopropylacetophenone

CAS Number: 645-13-6
MDL Number: MFCD00048297
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 238
Specific Gravity (20/20): 0.97

Formula: C₁₁H₁₄O
Boiling Pt: 238 °C
Density: 0.97 (20 °C)
MDL Number: MFCD00048297
CAS Number: 645-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=889c0bec97ae0a3d93305cf88af1c12345331359&vid=889c0bec97ae0a3d93305cf88af1c12345331359&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR