Methyl-5-iodsalicylate ≥98.0%

Supplier: TCI America

I0539-5G I0539-25G
TCI0539-005GEA 90.06 CAD
TCI0539-005G TCI0539-025G
Methyl-5-iodsalicylate ≥98.0%
Methyl 5-iodsalicylate

CAS Number: 4068-75-1
MDL Number: MFCD00016462
Molecular Formula: C8H7IO3
Molecular Weight: 278.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77


Other Product Names / Synonyms: 2-Hydroxy-5-iodobenzoic Acid Methyl Ester, Methyl 2-Hydroxy-5-iodobenzoate, 5-Iodosalicylic Acid Methyl Ester

Formula: C₈H₇IO₃
Melting Pt: 77 °C
MDL Number: MFCD00016462
CAS Number: 4068-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3329df3bcabd4494fce9378ca92c32bffdf4de2&vid=f3329df3bcabd4494fce9378ca92c32bffdf4de2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR