5-Iodo-2,4-dimethoxypyrimidine ≥98.0%

Supplier: TCI America

I0531-5G I0531-1G
TCI0531-005GEA 486.64 CAD
TCI0531-005G TCI0531-001G
5-Iodo-2,4-dimethoxypyrimidine ≥98.0%
5-Iodo-2,4-dimethoxypyrimidine

CAS Number: 52522-99-3
MDL Number: MFCD00090865
Molecular Formula: C6H7IN2O2
Molecular Weight: 266.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72

Formula: C₆H₇IN₂O₂
MW: 266.04 g/mol
Melting Pt: 72 °C
MDL Number: MFCD00090865
CAS Number: 52522-99-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c433f79fd119a450b31fa41fcc0da60a261d93c9&vid=c433f79fd119a450b31fa41fcc0da60a261d93c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR