L-Isoleucine methyl ester hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
H-Ile-OMe·HCl, H-Ile-OMe.HCl
I0522-1G
I0522-5G
TCI0522-1GEA
26.86
CAD
TCI0522-1G
TCI0522-005G
L-Isoleucine methyl ester hydrochloride ≥98.0%
L-Isoleucine methyl ester hydrochloride
CAS Number: 18598-74-8
MDL Number: MFCD00038911
Molecular Formula: C7H15NO2
Molecular Weight: 181.66
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 98
Specific rotation [a]20/D: 26.5 deg (C=2, H2O)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Ile-OMe.HCl
Formula:
C₇H₁₅NO₂ MW: 181.66 g/mol Melting Pt: 98 °C |
MDL Number:
MFCD00038911 CAS Number: 18598-74-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...