Di-tert-butyl iminodicarboxylate ≥95.0%

Supplier: TCI America

Synonyms: Di-(tert-butyl) imidodicarbonate

I0497-25G I0497-5G
TCI0497-25GEA 161.16 CAD
TCI0497-25G TCI0497-005G
Di-tert-butyl iminodicarboxylate ≥95.0%
Di-tert-butyl iminodicarboxylate

CAS Number: 51779-32-9
MDL Number: MFCD00043309
Molecular Formula: C10H19NO4
Molecular Weight: 217.27
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 117


Product Categories: Synthetic Organic Chemistry, Amination
Other Product Names / Synonyms: tert-Butyl Iminodicarboxylate, Iminodicarboxylic Acid Di-tert-butyl Ester

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00043309
CAS Number: 51779-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ffe91255f8930ef9bb2f5d37833ccf39b7631b1&vid=4ffe91255f8930ef9bb2f5d37833ccf39b7631b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR