Dimethyl-(-)-2,3-O-isopropylidene-L-tartrate ≥95.0%

Supplier: TCI America

I0447-5G I0447-25G
TCI0447-005GEA 83.74 CAD
TCI0447-005G TCI0447-25G
Dimethyl-(-)-2,3-O-isopropylidene-L-tartrate ≥95.0%
Dimethyl-(-)-2,3-O-isopropylidene-L-tartrate

CAS Number: 37031-29-1
MDL Number: MFCD00066218
Molecular Formula: C9H14O6
Molecular Weight: 218.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 139
Flash Point (°C): 110
Specific Gravity (20/20): 1.19
Specific rotation [a]20/D: -54 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: Dimethyl (4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylate, (4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylic Acid Dimethyl Ester, (-)-2,3-O-Isopropylidene-L-tartaric Acid Dimethyl Ester

Formula: C₉H₁₄O₆
Boiling Pt: 139 °C
Density: 1.19 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00066218
CAS Number: 37031-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c34cf5cc6648de09f2ffbb4989b3b6d9bbecbb9&vid=6c34cf5cc6648de09f2ffbb4989b3b6d9bbecbb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR