2-(4-Isobutylphenyl)propanoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (±)-Ibuprofen, [(±)-2-(4-Isobutylphenyl)-propionic Acid, Ibuprofen

I0415-500G I0415-25G I0415-100G
TCI0415-500GEA 867.69 CAD
TCI0415-500G TCI0415-025G TCI0415-100G
2-(4-Isobutylphenyl)propanoic acid ≥98.0% (by GC, titration analysis)
2-(4-Isobutylphenyl)propanoic acid

CAS Number: 15687-27-1
MDL Number: MFCD00010393
Molecular Formula: C13H18O2
Molecular Weight: 206.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 157
Melting point (°C): 76


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: Ibuprofen, 4-Isobutyl-alpha-methylphenylacetic Acid

Formula: C₁₃H₁₈O₂
MW: 206.3 g/mol
Boiling Pt: 157 °C
Melting Pt: 76 °C
MDL Number: MFCD00010393
CAS Number: 15687-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8674a1b68855144796e435b6140674e03f1ea4ac&vid=8674a1b68855144796e435b6140674e03f1ea4ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR