Iminostilbene ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

I0414-25G I0414-5G
TCI0414-025GEA 194.34 CAD
TCI0414-025G TCI0414-5G
Iminostilbene ≥98.0% (by GC, total nitrogen)
Iminostilbene

CAS Number: 256-96-2
MDL Number: MFCD00005071
Molecular Formula: C14H11N
Molecular Weight: 193.25
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 199


Other Product Names / Synonyms: 5H-Dibenzo[b,f]azepine

Formula: C₁₄H₁₁N
Melting Pt: 199 °C
MDL Number: MFCD00005071
CAS Number: 256-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8bd049aace7d8b1d0193aac6c328a0319cb59b5&vid=f8bd049aace7d8b1d0193aac6c328a0319cb59b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR