Isosorbide ≥98.0%

Supplier: TCI America

Synonyms: D-Isosorbide, (3R,3aR,6S,6aR)-Hexahydrofuro[3,2-b]furan-3,6-diol

I0407-25G I0407-500G I0407-100G
TCI0407-025GEA 45.82 CAD
TCI0407-025G TCI0407-500G TCI0407-100G
Isosorbide ≥98.0%
Isosorbide

CAS Number: 652-67-5
MDL Number: MFCD00064827
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 175
Melting point (°C): 61
Specific rotation [a]20/D: 45 deg (C=5, H2O)


Product Categories: Bioscience, Sugars (Others), Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 1,4:3,6-Dianhydroglucitol, 1,4:3,6-Dianhydrosorbitol

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 175 °C
Melting Pt: 61 °C
MDL Number: MFCD00064827
CAS Number: 652-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54ffe893107ed94d94f9512127ee666a3af10e0&vid=f54ffe893107ed94d94f9512127ee666a3af10e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR