3-Hydroxy-2,2,4-trimethylpentyl isobutyrate 60.0% (contains ca. 40% 2,2,4-Trimethyl-1,3-pentanediol 3-Monoisobutyrate)

Supplier: TCI America

Synonyms: 2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate, 2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate, Isobutyric Acid 3-Hydroxy-2,2,4-trimethylpentyl Ester, 2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate)

I0405-25ML I0405-500ML
TCI0405-025MLEA 26.86 CAD
TCI0405-025ML TCI0405-500ML
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate 60.0% (contains ca. 40% 2,2,4-Trimethyl-1,3-pentanediol 3-Monoisobutyrate)
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate

CAS Number: 25265-77-4
MDL Number: MFCD00148967
Molecular Formula: C12H24O3
Molecular Weight: 216.32
Purity/Analysis Method: >60.0% (GC)
Form: Clear Liquid
Boiling point (°C): 253
Melting point (°C): -50
Flash Point (°C): 123
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Isobutyric Acid 3-Hydroxy-2,2,4-trimethylpentyl Ester, 2,2,4-Trimethyl-1,3-pentanediol 1-Monoisobutyrate

Formula: C₁₂H₂₄O₃
MW: 216.32 g/mol
Boiling Pt: 253 °C
Melting Pt: –50 °C
Density: 0.95 (20 °C)
Flash Pt: 123 °C
MDL Number: MFCD00148967
CAS Number: 25265-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b454b9efc21ad63efb93b84e7918bc4c45cee018&vid=af923ffec39cff68c984ca4c2e1b59a0ceea1e3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 60.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR