Sodium-2-naphthalenesulfonate ≥98.0%
Supplier: TCI America
Synonyms:
2-Naphthalenesulphonic acid Sodium Salt, 2-Naphthalenesulfonic acid Sodium Salt, Sodium 2-naphthalenesulphonate, Sodium 2-naphthalenesulfonate
I0353-5G
I0353-25G
TCI0353-005GEA
55.3
CAD
TCI0353-005G
TCI0353-025G
Sodium-2-naphthalenesulfonate ≥98.0%
Sodium-2-naphthalenesulfonate
CAS Number: 532-02-5
MDL Number: MFCD00064186
Molecular Formula: C10H8O3S
Molecular Weight: 230.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 2-Naphthalenesulfonic Acid Sodium Salt
Formula:
C₁₀H₈O₃S MW: 230.21 g/mol |
MDL Number:
MFCD00064186 CAS Number: 532-02-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...