1-Nonanesulfonic acid sodium salt ≥98.0%

Supplier: TCI America

Synonyms: Sodium 1-nonanesulfonate, Sodium nonane-1-sulfonate, Sodium 1-nonanesulphonate, Sodium nonane-1-sulphonate

I0347-25G I0347-5G
TCI0347-025GEA 307.47 CAD
TCI0347-025G TCI0347-005G
1-Nonanesulfonic acid sodium salt ≥98.0%
1-Nonanesulphonic acid sodium salt

[Reagent for Ion-Pair Chromatography]
CAS Number: 35192-74-6
MDL Number: MFCD00025010
Molecular Formula: C9H20O3S
Molecular Weight: 230.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: IPC-ALKS-9, 1-Nonanesulfonic Acid Sodium Salt

Formula: C₉H₁₉NaO₃S
MDL Number: MFCD00025010
CAS Number: 35192-74-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f561f760fda6821e1496b45e8ea07f0789cebaf2&vid=c13f9a0dd306fb6b3189881b6d5865d73b638060&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR