Sodium-1-propanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Propanesulphonic acid sodium salt, 1-Propanesulphonic acid Sodium Salt, Propanesulfonic acid Sodium Salt, Propanesulphonic acid Sodium Salt, Propanesulfonic acid sodium salt, 1-Propanesulfonic acid Sodium Salt, IPC-ALKS-3

I0341-25G I0341-5G
TCI0341-025GEA 401.32 CAD
TCI0341-025G TCI0341-005G
Sodium-1-propanesulfonate ≥98.0% (by titrimetric analysis)
Sodium-1-propanesulfonate

[Reagent for Ion-Pair Chromatography]
CAS Number: 14533-63-2
MDL Number: MFCD00062546
Molecular Formula: C3H8O3S
Molecular Weight: 146.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 255


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: IPC-ALKS-3, 1-Propanesulfonic Acid Sodium Salt

Formula: C₃H₈O₃S
MW: 146.14 g/mol
Melting Pt: 255 °C
MDL Number: MFCD00062546
CAS Number: 14533-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1b9a78e9df28d8e459e2f74f3f2c2eb1d254511&vid=a1b9a78e9df28d8e459e2f74f3f2c2eb1d254511&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR