Isobornyl acetate ≥90.0%
Supplier: TCI America
I0306-500ML
I0306-25ML
TCI0306-500MLEA
82.16
CAD
TCI0306-500ML
TCI0306-025ML
Isobornyl acetate ≥90.0%
Isobornyl acetate
CAS Number: 125-12-2
MDL Number: MFCD00135943
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Melting point (°C): 7
Flash Point (°C): 100
Specific Gravity (20/20): 0.99
Specific rotation [a]20/D: 0.8 deg (neat)
Formula:
C₁₂H₂₀O₂ Boiling Pt: 106 °C Melting Pt: 7 °C Density: 0.99 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00135943 CAS Number: 125-12-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...