Ethyl-4-piperidinecarboxylate ≥98.0%
Supplier: TCI America
I0294-25ML
I0294-500ML
I0294-100ML
TCI0294-025MLEA
57.88
CAD
TCI0294-025ML
TCI0294-500ML
TCI0294-100ML
Ethyl-4-piperidinecarboxylate ≥98.0%
Ethyl 4-piperidinecarboxylate
CAS Number: 1126-09-6
MDL Number: MFCD00006003
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 80
Specific Gravity (20/20): 1.02
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl Isonipecotate, Isonipecotic Acid Ethyl Ester, 4-Piperidinecarboxylic Acid Ethyl Ester
Formula:
C₈H₁₅NO₂ Boiling Pt: 105 °C Density: 1.02 (20 °C) Flash Pt: 80 °C |
MDL Number:
MFCD00006003 CAS Number: 1126-09-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...