(±)-Isoborneol ≥90.0%

Supplier: TCI America

I0275-25G I0275-500G
TCI0275-025GEA 31.6 CAD
TCI0275-025G TCI0275-500G
(±)-Isoborneol ≥90.0%
(±)-Isoborneol

CAS Number: 124-76-5
MDL Number: MFCD00074821
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Melting point (°C): 211

Formula: C₁₀H₁₈O
Melting Pt: 211 °C
MDL Number: MFCD00074821
CAS Number: 124-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c06e74c29aefd0fe0391d23d8eba80e429dacbf&vid=7c06e74c29aefd0fe0391d23d8eba80e429dacbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR