Isonipecotamide ≥98.0%

Supplier: TCI America

I0255-250G I0255-25G
TCI0255-250GEA 623.15 CAD
TCI0255-250G TCI0255-025G
Isonipecotamide ≥98.0%
Isonipecotamide

CAS Number: 39546-32-2
MDL Number: MFCD00038012
Molecular Formula: C6H12N2O
Molecular Weight: 128.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 150
Flash Point (°C): 90


Other Product Names / Synonyms: 4-Piperidinecarboxamide

Formula: C₆H₁₂N₂O
Melting Pt: 150 °C
Flash Pt: 90 °C
MDL Number: MFCD00038012
CAS Number: 39546-32-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f60a9a37053de0d86ebd11a56713644db762cca4&vid=f60a9a37053de0d86ebd11a56713644db762cca4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR