Diethyl isobutylmalonate ≥98.0%

Supplier: TCI America

Synonyms: Isobutylmalonic Acid Diethyl Ester

I0251-500ML I0251-25ML
TCI0251-500MLEA 991.77 CAD
TCI0251-500ML TCI0251-025ML
Diethyl isobutylmalonate ≥98.0%
Diethyl isobutylmalonate

CAS Number: 10203-58-4
MDL Number: MFCD00026869
Molecular Formula: C11H20O4
Molecular Weight: 216.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 225
Flash Point (°C): 100
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Isobutylmalonic Acid Diethyl Ester

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 225 °C
Density: 0.97 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00026869
CAS Number: 10203-58-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa5cb059229a2854d94c4e32c110d9a7572bdbb0&vid=aa5cb059229a2854d94c4e32c110d9a7572bdbb0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR