2-(1H-Imidazol-5-yl)acetic acid hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Imidazoleacetic acid hydrochloride, Imidazole-4-acetic acid monohydrochloride

I0229-1G I0229-5G
TCI0229-001GEA 273.34 CAD
TCI0229-001G TCI0229-5G
2-(1H-Imidazol-5-yl)acetic acid hydrochloride ≥98.0% (by titrimetric analysis)
2-(1H-Imidazol-5-yl)acetic acid hydrochloride

CAS Number: 3251-69-2
MDL Number: MFCD00012698
Molecular Formula: C5H6N2O2
Molecular Weight: 162.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 225


Other Product Names / Synonyms: 4-Carboxymethylimidazole Hydrochloride

Formula: C₅H₇ClN₂O₂
MW: 162.58 g/mol
Melting Pt: 225 °C
MDL Number: MFCD00012698
CAS Number: 3251-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0dacad7412b2bd11a20f406786369687ba16a0da&vid=0dacad7412b2bd11a20f406786369687ba16a0da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR