3,3'-Iminodipropionitrile ≥98.0%

Supplier: TCI America

I0010-25ML I0010-100ML
TCI0010-025MLEA 147.51 CAD
TCI0010-025ML TCI0010-100ML
3,3'-Iminodipropionitrile ≥98.0%
3,3'-Iminodipropionitrile

CAS Number: 111-94-4
MDL Number: MFCD00001956
Molecular Formula: C6H9N3
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 178
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Bis(2-cyanoethyl)amine

Formula: C₆H₉N₃
Boiling Pt: 205 °C
Density: 1.03 (20 °C)
Flash Pt: 178 °C
MDL Number: MFCD00001956
CAS Number: 111-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce43f413ebfb7cc400c455a821fbccacc33f1079&vid=ce43f413ebfb7cc400c455a821fbccacc33f1079&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR