1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulfonate ≥98.0%

Supplier: TCI America

H1281-5G H1281-25G
TCH1281-5GEA 135.88 CAD
TCH1281-5G TCH1281-25G
1,1,1,3,3,3-Hexafluoroisopropyltoluene-4-sulfonate ≥98.0%
1,1,1,3,3,3-Hexafluoroisopropyl toluene-4-sulphonate

CAS Number: 67674-48-0
MDL Number: MFCD00039262
Molecular Formula: C10H8F6O3S
Molecular Weight: 322.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 134
Melting point (°C): 45


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Fluorous Compounds
Other Product Names / Synonyms: p-Toluenesulfonic Acid 1,1,1,3,3,3-Hexafluoroisopropyl Ester, 1,1,1,3,3,3-Hexafluoroisopropyl Tosylate

Formula: C₁₀H₈F₆O₃S
Boiling Pt: 134 °C
Melting Pt: 45 °C
MDL Number: MFCD00039262
CAS Number: 67674-48-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e141aba057a29a1c6f027c4c6747d589e050aa85&vid=e141aba057a29a1c6f027c4c6747d589e050aa85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR