Azetidin-3-ol hydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI America
H1264-5G
H1264-1G
TCH1264-5GEA
203.35
CAD
TCH1264-5G
TCH1264-1G
Azetidin-3-ol hydrochloride ≥98.0% (by total nitrogen basis)
Azetidin-3-ol hydrochloride
CAS Number: 18621-18-6
MDL Number: MFCD02683887
Molecular Formula: C3H7NO
Molecular Weight: 109.55
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 89
Other Product Names / Synonyms: 3-Azetidinol Hydrochloride
Formula:
C₃H₈ClNO Melting Pt: 89 °C |
MDL Number:
MFCD02683887 CAS Number: 18621-18-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...