5-Isoquinolinol ≥96.0%

Supplier: TCI America

Synonyms: 5-Hydroxyisoquinoline

H1259-25G H1259-5G
TCH1259-25GEA 1153.72 CAD
TCH1259-25G TCH1259-5G
5-Isoquinolinol ≥96.0%
5-Isoquinolinol

CAS Number: 2439-04-5
MDL Number: MFCD00006906
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 220


Other Product Names / Synonyms: 5-Isoquinolinol

Formula: C₉H₇NO
MW: 145.16
Melting Pt: 220 °C
MDL Number: MFCD00006906
CAS Number: 2439-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a555d13753483697a6fbf6cf9af0bf2711fd1468&vid=a555d13753483697a6fbf6cf9af0bf2711fd1468&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR