3-Hydroxy-1-adamantane carboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Hydroxyadamantane-1-carboxylic acid, 3-Hydroxy-1-adamantanecarboxylic acid

H1255-1G H1255-5G H1255-25G
TCH1255-1GEA 34.76 CAD
TCH1255-1G TCH1255-5G TCH1255-25G
3-Hydroxy-1-adamantane carboxylic acid ≥97.0% (by GC, titration analysis)
3-Hydroxy-1-adamantane carboxylic acid

CAS Number: 42711-75-1
MDL Number: MFCD00193929
Molecular Formula: C11H16O3
Molecular Weight: 196.25
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 203

Formula: C₁₁H₁₆O₃
MW: 196.25 g/mol
Melting Pt: 203 °C
MDL Number: MFCD00193929
CAS Number: 42711-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f93aa489a85084d911bdc117c0b9352448d2cdf&vid=6f93aa489a85084d911bdc117c0b9352448d2cdf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR